Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-25
Entrez Gene ID
174412
Gene Name
Zinc metalloproteinase nas-25
Gene Aliases
CELE_F46C5.3
Chromosome Location
Chr.II: 8826069 - 8828215 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063479.5 NP_495880.1 9 - 10 1125 92

View Full Product Details