Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-78
Entrez Gene ID
189274
Gene Name
COLlagen
Gene Aliases
CELE_W07A12.5
Chromosome Location
Chr.II: 9152531 - 9154336 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377833.2 NP_001364618.1 3 - 4 459 104

View Full Product Details