Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab-39
Entrez Gene ID
183941
Gene Name
RAB family
Gene Aliases
CELE_D2013.1
Chromosome Location
Chr.II: 9321858 - 9322920 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063583.4 NP_495984.2 2 - 3 442 94

View Full Product Details