Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-3
Entrez Gene ID
191641
Gene Name
protein-coding
Gene Aliases
CELE_R134.1
Chromosome Location
Chr.II: 9502757 - 9507281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063636.1 NP_496037.1 8 - 9 1352 62

View Full Product Details