Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sqv-8
- Entrez Gene ID
-
174517
- Gene Name
- Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase sqv-8
- Gene Aliases
-
CELE_ZK1307.5
- Chromosome Location
-
Chr.II: 9647878 - 9649367 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063675.6 |
NP_496076.1 |
2 - 3 |
307 |
110 |
GenBank mRNA |
AJ005864.1 |
- |
2 - 3 |
303 |
110 |
- Gene Symbol
- ZK1307.9
- Entrez Gene ID
-
174518
- Gene Name
- Coiled-coil domain-containing protein 130 homolog
- Gene Aliases
-
CELE_ZK1307.9
- Chromosome Location
-
Chr.II: 9647894 - 9650963 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063676.6 |
NP_496077.1 |
2 - 3 |
291 |
110 |
View Full Product Details