Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aqp-10
Entrez Gene ID
174537
Gene Name
Putative aquaporin-10
Gene Aliases
CELE_ZK1321.3
Chromosome Location
Chr.II: 9768357 - 9769847 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063704.6 NP_496105.1 2 - 3 189 85

View Full Product Details