Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A6
Entrez Gene ID
188357
Gene Name
Putative cytochrome P450 CYP13A6
Gene Aliases
CELE_T10B9.3
Chromosome Location
Chr.II: 9800758 - 9802821 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063712.4 NP_496113.1 3 - 4 708 75

View Full Product Details