Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psf-1
Entrez Gene ID
187885
Gene Name
Probable DNA replication complex GINS protein PSF1
Gene Aliases
CELE_R53.6
Chromosome Location
Chr.II: 9968673 - 9969631 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063752.5 NP_496153.2 2 - 3 168 91

View Full Product Details