Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cut-4
Entrez Gene ID
184037
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_E04D5.3
Chromosome Location
Chr.II: 10436135 - 10440087 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063841.3 NP_496242.2 5 - 6 1089 73

View Full Product Details