Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ggr-1
Entrez Gene ID
191636
Gene Name
Glycine receptor subunit beta-type 4
Gene Aliases
CELE_C09G5.1
Chromosome Location
Chr.II: 10695013 - 10702555 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063905.4 NP_496306.2 7 - 8 1409 100

View Full Product Details