Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-12
Entrez Gene ID
174655
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_C52A11.2
Chromosome Location
Chr.II: 10729804 - 10731652 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063913.2 NP_496314.1 3 - 4 480 144

View Full Product Details