Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-14
Entrez Gene ID
174662
Gene Name
SET domain-containing protein 14
Gene Aliases
CELE_R06F6.4
Chromosome Location
Chr.II: 10793575 - 10796709 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063922.6 NP_496323.1 4 - 5 908 101

View Full Product Details