Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egg-3
Entrez Gene ID
174677
Gene Name
Protein tyrosine phosphatase-like protein egg-3
Gene Aliases
CELE_F44F4.2
Chromosome Location
Chr.II: 10882407 - 10884441 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383944.1 2 - 3 364 84

View Full Product Details