Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
raga-1
Entrez Gene ID
174726
Gene Name
RAs-related GTP-binding protein A
Gene Aliases
CELE_T24F1.1
Chromosome Location
Chr.II: 11297698 - 11302832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383949.1 12 - 13 2854 107

View Full Product Details