Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-6
Entrez Gene ID
174743
Gene Name
V-type proton ATPase subunit a
Gene Aliases
CELE_VW02B12L.1
Chromosome Location
Chr.II: 11435210 - 11438452 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064035.9 NP_496436.1 3 - 4 1754 78
GenBank mRNA AB055111.1 - 3 - 4 1776 78

View Full Product Details