Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ooc-3
Entrez Gene ID
174759
Gene Name
protein-coding
Gene Aliases
CELE_B0334.11
Chromosome Location
Chr.II: 11493159 - 11495321 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064055.4 NP_496456.1 1 - 2 182 81

View Full Product Details