Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-84
Entrez Gene ID
174824
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_K12D12.3
Chromosome Location
Chr.II: 11868534 - 11869688 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064134.6 NP_496535.1 1 - 2 152 84

View Full Product Details