Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-29
Entrez Gene ID
189454
Gene Name
protein-coding
Gene Aliases
CELE_Y17G7B.6
Chromosome Location
Chr.II: 12009072 - 12023832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267402.3 NP_001254331.1 9 - 10 1048 68
NM_001267403.3 NP_001254332.1 9 - 10 1075 68
NM_001267404.4 NP_001254333.1 7 - 8 826 68
NM_001267405.2 NP_001254334.1 5 - 6 579 68

View Full Product Details