Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
set-24
Entrez Gene ID
3565594
Gene Name
SET (trithorax/polycomb) domain containing
Gene Aliases
CELE_Y43F11A.5
Chromosome Location
Chr.II: 12114971 - 12130970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064178.6 NP_496579.3 3 - 4 886 142

View Full Product Details