Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqn-87
Entrez Gene ID
174861
Gene Name
protein-coding
Gene Aliases
CELE_Y57A10A.18
Chromosome Location
Chr.II: 12209095 - 12225086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267406.4 NP_001254335.1 13 - 14 4310 80
NM_001267407.4 NP_001254336.1 13 - 14 4450 80
NM_001381626.2 NP_001368162.1 13 - 14 4459 80
NM_064193.4 NP_496594.2 14 - 15 4489 80

View Full Product Details