Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-56
Entrez Gene ID
183669
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_C50E10.8
Chromosome Location
Chr.II: 12324459 - 12327565 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064216.2 NP_496617.1 3 - 4 1190 86

View Full Product Details