Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-8
Entrez Gene ID
174893
Gene Name
C-type LECtin
Gene Aliases
CELE_Y38E10A.4
Chromosome Location
Chr.II: 12575691 - 12578061 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064286.6 NP_496687.2 3 - 4 812 71

View Full Product Details