Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-14
Entrez Gene ID
185262
Gene Name
Serpentine Receptor, class A (alpha)
Gene Aliases
CELE_F35C5.2
Chromosome Location
Chr.II: 12885272 - 12888608 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064337.7 NP_496738.2 4 - 5 668 77

View Full Product Details