Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zyx-1
Entrez Gene ID
174951
Gene Name
protein-coding
Gene Aliases
CELE_F42G4.3
Chromosome Location
Chr.II: 13056814 - 13071530 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267435.3 NP_001254364.1 1 - 2 45 61
NM_064375.8 NP_496776.1 2 - 3 297 61

View Full Product Details