Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-45
Entrez Gene ID
185115
Gene Name
BTB and MATH domain-containing protein 45
Gene Aliases
CELE_F29C12.5
Chromosome Location
Chr.II: 13123020 - 13125189 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064387.3 NP_496788.1 4 - 5 809 80

View Full Product Details