Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hpr-17
Entrez Gene ID
174958
Gene Name
Homolog of S. Pombe Rad
Gene Aliases
CELE_F32A11.2
Chromosome Location
Chr.II: 13153461 - 13157047 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064392.6 NP_496793.1 3 - 4 534 108
GenBank mRNA AF076840.1 - 3 - 4 521 108

View Full Product Details