Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gst-14
Entrez Gene ID
185409
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_F37B1.3
Chromosome Location
Chr.II: 13614781 - 13616100 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064460.2 NP_496861.1 1 - 2 143 66

View Full Product Details