Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nurf-1
Entrez Gene ID
175098
Gene Name
protein-coding
Gene Aliases
CELE_F26H11.2
Chromosome Location
Chr.II: 14390713 - 14407911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026946.4 NP_001022117.2 6 - 7 1896 95
NM_001267499.2 NP_001254428.1 6 - 7 1900 95
NM_001267500.3 NP_001254429.1 6 - 7 1901 95
NM_001313609.3 NP_001300538.1 5 - 6 1531 95
NM_001313610.3 NP_001300539.1 5 - 6 1531 95
NM_001313611.3 NP_001300540.1 6 - 7 1894 95
NM_001377899.3 NP_001364539.1 6 - 7 1900 95
NM_001381656.2 NP_001366940.1 6 - 7 1901 95
NM_001383991.1 7 - 8 1811 95
NM_001383992.1 6 - 7 1880 95
NM_064593.4 NP_496994.3 6 - 7 1901 95
NM_064594.9 NP_496995.3 6 - 7 1900 95

View Full Product Details