Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nurf-1
Entrez Gene ID
175098
Gene Name
protein-coding
Gene Aliases
CELE_F26H11.2
Chromosome Location
Chr.II: 14390713 - 14407911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026946.4 NP_001022117.2 23 - 24 6099 67
NM_001026948.5 NP_001022119.1 4 - 5 844 67
NM_001026949.5 NP_001022120.1 4 - 5 1158 67
NM_001313609.3 NP_001300538.1 22 - 23 5740 67
NM_001313610.3 NP_001300539.1 22 - 23 5734 67
NM_001313611.3 NP_001300540.1 23 - 24 6103 67
NM_001313612.3 NP_001300541.1 7 - 8 1219 67
NM_001313613.3 NP_001300542.1 4 - 5 871 67
NM_001377899.3 NP_001364539.1 23 - 24 6118 67
NM_001381656.2 NP_001366940.1 23 - 24 6110 67
NM_064594.9 NP_496995.3 23 - 24 6112 67

View Full Product Details