Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atgp-2
Entrez Gene ID
175125
Gene Name
protein-coding
Gene Aliases
CELE_C38C6.2
Chromosome Location
Chr.II: 14640089 - 14644083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267506.3 NP_001254435.1 5 - 6 1097 69
NM_001267507.3 NP_001254436.1 4 - 5 956 69

View Full Product Details