Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-20
Entrez Gene ID
175165
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_Y53F4B.28
Chromosome Location
Chr.II: 15158174 - 15163614 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064713.5 NP_497114.1 10 - 11 2050 151

View Full Product Details