Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mat-3
Entrez Gene ID
175205
Gene Name
Cell division cycle protein 23 homolog
Gene Aliases
CELE_F10C5.1
Chromosome Location
Chr.III: 475454 - 482558 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064802.6 NP_497203.1 5 - 6 1229 62

View Full Product Details