Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptl-1
Entrez Gene ID
175230
Gene Name
protein-coding
Gene Aliases
CELE_F42G9.9
Chromosome Location
Chr.III: 763810 - 771391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027405.6 NP_001022576.1 8 - 9 1351 91
NM_001384010.1 5 - 6 1301 91
GenBank mRNA U67966.1 - 7 - 8 1297 91

View Full Product Details