Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trp-2
Entrez Gene ID
192056
Gene Name
protein-coding
Gene Aliases
CELE_R06B10.4
Chromosome Location
Chr.III: 983261 - 1001931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027532.5 NP_001022703.1 8 - 9 1281 76
NM_064910.8 NP_497311.2 14 - 15 2540 76
GenBank mRNA AJ276028.1 - 13 - 14 2370 76

View Full Product Details