Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tig-3
Entrez Gene ID
266879
Gene Name
TGF_BETA_2 domain-containing protein
Gene Aliases
CELE_Y46E12BL.1
Chromosome Location
Chr.II: 15244472 - 15251792 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064917.6 NP_497318.2 4 - 5 765 66

View Full Product Details