Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chin-1
Entrez Gene ID
175268
Gene Name
CHImaeriN (Rac-GTPase-activating protein) homolog
Gene Aliases
CELE_BE0003N10.2
Chromosome Location
Chr.III: 1031318 - 1037212 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064922.7 NP_497323.3 1 - 2 33 93

View Full Product Details