Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smr-1
Entrez Gene ID
175281
Gene Name
SMN (Survival of Motor Neuron protein) Related
Gene Aliases
CELE_Y71D11A.2
Chromosome Location
Chr.III: 1143172 - 1145222 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027761.7 NP_001022932.1 1 - 2 141 88

View Full Product Details