Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-28
Entrez Gene ID
191033
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y119D3A.2
Chromosome Location
Chr.III: 1297904 - 1299484 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064975.1 NP_497376.1 2 - 3 675 139

View Full Product Details