Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-74
Entrez Gene ID
189515
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y22D7AR.9
Chromosome Location
Chr.III: 1696952 - 1698986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065045.7 NP_497446.3 1 - 2 143 74

View Full Product Details