Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppk-2
Entrez Gene ID
175344
Gene Name
PIPK domain-containing protein
Gene Aliases
CELE_Y48G9A.8
Chromosome Location
Chr.III: 2102295 - 2107497 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065099.5 NP_497500.1 5 - 6 1184 73

View Full Product Details