Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mdt-19
Entrez Gene ID
175378
Gene Name
Mediator of RNA polymerase II transcription subunit 19
Gene Aliases
CELE_Y71H2B.6
Chromosome Location
Chr.III: 2598053 - 2603299 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065186.4 NP_497587.1 1 - 2 97 112

View Full Product Details