Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mdt-19
- Entrez Gene ID
-
175378
- Gene Name
- Mediator of RNA polymerase II transcription subunit 19
- Gene Aliases
-
CELE_Y71H2B.6
- Chromosome Location
-
Chr.III: 2598053 - 2603299 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065186.4 |
NP_497587.1 |
2 - 3 |
372 |
65 |
View Full Product Details