Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdh-1
Entrez Gene ID
187783
Gene Name
CaDHerin family
Gene Aliases
CELE_R10F2.1
Chromosome Location
Chr.III: 2910906 - 2935961 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065240.3 NP_497641.2 2 - 3 448 70

View Full Product Details