The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267937.3 | NP_001254866.1 | 5 - 6 | 741 | 149 |
NM_001267938.2 | NP_001254867.1 | 4 - 5 | 333 | 149 |