Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mlc-4
Entrez Gene ID
175440
Gene Name
Myosin regulatory light chain
Gene Aliases
CELE_C56G7.1
Chromosome Location
Chr.III: 3392514 - 3393795 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065299.6 NP_497700.1 1 - 2 113 64

View Full Product Details