Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cri-3
Entrez Gene ID
175441
Gene Name
Conserved regulator of innate immunity protein 3
Gene Aliases
CELE_F59A2.3
Chromosome Location
Chr.III: 3396355 - 3400606 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384052.1 4 - 5 652 82

View Full Product Details