Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
har-1
Entrez Gene ID
175529
Gene Name
Hemiasterlin resistant protein 1
Gene Aliases
CELE_C16C10.11
Chromosome Location
Chr.III: 4152054 - 4152915 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065425.4 NP_497826.1 1 - 2 108 74

View Full Product Details