Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sel-5
Entrez Gene ID
175599
Gene Name
protein-coding
Gene Aliases
CELE_F35G12.3
Chromosome Location
Chr.III: 4571748 - 4578437 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027391.8 NP_001022562.1 14 - 15 2344 138
GenBank mRNA AF256466.1 - 14 - 15 2334 138

View Full Product Details