Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
emb-5
Entrez Gene ID
175621
Gene Name
Suppressor of Ty 6 homolog
Gene Aliases
CELE_T04A8.14
Chromosome Location
Chr.III: 4715293 - 4720670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065568.4 NP_497969.1 4 - 5 1320 65

View Full Product Details