Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- coq-8
- Entrez Gene ID
-
175647
- Gene Name
- Atypical kinase coq-8, mitochondrial
- Gene Aliases
-
CELE_C35D10.4
- Chromosome Location
-
Chr.III: 4862354 - 4866953 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_065613.9 |
NP_498014.2 |
6 - 7 |
2042 |
119 |
- Gene Symbol
- C35D10.17
- Entrez Gene ID
-
259504
- Gene Name
- COX assembly mitochondrial protein 2 homolog
- Gene Aliases
-
CELE_C35D10.17
- Chromosome Location
-
Chr.III: 4863421 - 4867469 on Build WBcel235
View Full Product Details